2,4-Difluorbenzoylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,4-Difluorbenzoesäurechlorid

A17920.06 A17920.14
A17920.06EA 45 EUR
A17920.06 A17920.14
2,4-Difluorbenzoylchlorid ≥98%
2,4-Difluorbenzoylchlorid

Purity: 98% CAS No.: 72482-64-5 Molecular Formula: C7H3ClF2O Formula Weight: 176.55 Flash Point: 82°(179°F) Refractive Index: 1.5145 Boiling Point: 96-98°/15mm Beilstein Reference: 4800566 UN No.: 3265 MDL No.: MFCD00000658

Formel: C₇H₃ClF₂O
Molecular Weight: 176,55 g/mol
Siedepunkt: 96…98 °C (15 mmHg)
Dichte: 1,437
Flammpunkt: 82 °C (179 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000658
CAS-Nummer: 72482-64-5
EINECS: 276-690-4
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR