2,4-Difluorbenzoylchlorid

Lieferant: Thermo Scientific

Synonyme: 2,4-Difluorbenzoesäurechlorid

TL00171.1 TL00171.10 TL00171.50
MAYBTL00171.1EA 0 EUR
MAYBTL00171.1 MAYBTL00171.10 MAYBTL00171.50
2,4-Difluorbenzoylchlorid
2,4-Difluorbenzoylchlorid
Formel: C₇H₃ClF₂O
Molecular Weight: 176,55 g/mol
Siedepunkt: 68 °C (1013 hPa)
Dichte: 1,42 g/cm³ (20 °C)
MDL: MFCD00000658
CAS-Nummer: 72482-64-5
EINECS: 276-690-4
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR