2,4-Difluorbenzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2,4-Difluorbenzoesäurechlorid
004789-25G
004789-100G
FLUO004789-25GEA
134
EUR
FLUO004789-25G
FLUO004789-100G
2,4-Difluorbenzoylchlorid
2,4-Difluorbenzoylchlorid
Formel:
C₇H₃ClF₂O Siedepunkt: 68 °C (1013 hPa) Dichte: 1,42 g/cm³ (20 °C) |
MDL:
MFCD00000658 CAS-Nummer: 72482-64-5 EINECS: 276-690-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...