2,6-Difluorbenzoylchlorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,6-Difluorbenzoesäurechlorid
A11286.22
A11286.06
A11286.14
A11286.22EA
187
EUR
A11286.22
A11286.06
A11286.14
2,6-Difluorbenzoylchlorid ≥98%
2,6-Difluorbenzoylchlorid
Purity: 98%
CAS No.: 18063-02-0
Molecular Formula: C7H3ClF2O
Formula Weight: 176.55
Flash Point: 72°(162°F)
Refractive Index: 1.4988
Boiling Point: 72-77°/13mm
Beilstein Reference: 639438
UN No.: 3265
MDL No.: MFCD00000659
Formel:
C₇H₃ClF₂O Molecular Weight: 176,55 g/mol Siedepunkt: 72…77 °C (13 mmHg) Dichte: 1,407 Flammpunkt: 72 °C (162 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000659 CAS-Nummer: 18063-02-0 EINECS: 241-971-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...