2,6-Difluorbenzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2,6-Difluorbenzoesäurechlorid
004790-25G
004790-100G
004790-250G
FLUO004790-25GEA
31.1
EUR
FLUO004790-25G
FLUO004790-100G
FLUO004790-250G
2,6-Difluorbenzoylchlorid
2,6-Difluorbenzoylchlorid
Formel:
C₇H₃ClF₂O Molecular Weight: 176,55 g/mol Siedepunkt: 72 °C (1013 hPa) Dichte: 1,4 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000659 CAS-Nummer: 18063-02-0 EINECS: 241-971-2 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...