3,4,5-Trifluorbenzoylchlorid
Lieferant: FLUOROCHEM
Synonyme:
3,4,5-Trifluorbenzoesäurechlorid
007497-1G
007497-5G
FLUO007497-1GEA
76.2
EUR
FLUO007497-1G
FLUO007497-5G
3,4,5-Trifluorbenzoylchlorid
3,4,5-Trifluorbenzoylchlorid
Formel:
C₇H₂ClF₃O Molecular Weight: 194.54 g/mol Siedepunkt: 186 Schmelzpunkt: 41 |
MDL:
MFCD00083526 CAS-Nummer: 177787-26-7 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...