3,5-Difluorbenzoylchlorid

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 3,5-Difluorbenzoesäurechlorid

002113-10G 002113-25G 002113-100G
FLUO002113-10GEA 36.9 EUR
FLUO002113-10G FLUO002113-25G FLUO002113-100G
3,5-Difluorbenzoylchlorid
3,5-Difluorbenzoylchlorid
Formel: C₇H₃ClF₂O
Molecular Weight: 176.55 g/mol
Dichte: 1.5 g/cm³ (20 °C)
MDL: MFCD00010309
CAS-Nummer: 129714-97-2
UN: 3265

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR