Pentafluorbenzoylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: Pentafluorbenzoesäurechlorid, Perfluorbenzoylchlorid

A12420.06 A12420.14 A12420.22
A12420.06EA 58.8 EUR
A12420.06 A12420.14 A12420.22
Pentafluorbenzoylchlorid ≥98%
Pentafluorbenzoylchlorid

Purity: 98% CAS No.: 2251-50-5 Molecular Formula: C7ClF5O Formula Weight: 230.52 Refractive Index: 1.4535 Boiling Point: 158-159° UN No.: 3265 MDL No.: MFCD00000657

Formel: C₇ClF₅O
Molecular Weight: 230,52 g/mol
Siedepunkt: 158…159 °C
Dichte: 1,668
Lagertemperatur: Raumtemperatur
MDL: MFCD00000657
CAS-Nummer: 2251-50-5
EINECS: 218-842-4
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR