2-(Trifluormethoxy)benzoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluoromethoxy)benzene-1-carbonyl chloride

007546-5G 007546-25G
FLUO007546-5GEA 51.8 EUR
FLUO007546-5G FLUO007546-25G
2-(Trifluormethoxy)benzoylchlorid
2-(Trifluormethoxy)benzoylchlorid
Formel: C₈H₄ClF₃O₂
Molecular Weight: 224.57 g/mol
Siedepunkt: 94…96 °C (15 mmHg)
MDL: MFCD00052330
CAS-Nummer: 162046-61-9
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR