3-(Trifluormethoxy)benzoylchlorid

Lieferant: FLUOROCHEM

007547-5G 007547-25G 007547-1G
FLUO007547-5GEA 0 EUR
FLUO007547-5G FLUO007547-25G FLUO007547-1G
3-(Trifluormethoxy)benzoylchlorid
3-(Trifluormethoxy)benzoylchlorid
Formel: C₈H₄ClF₃O₂
MDL: MFCD00061268
CAS-Nummer: 86270-03-3
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR