3-Fluor-4-methoxybenzoylchlorid

Lieferant: FLUOROCHEM

Synonyme: 3-Fluor-4-methoxybenzoesäurechlorid

013057-5G 013057-25G
FLUO013057-5GEA 42.9 EUR
FLUO013057-5G FLUO013057-25G
3-Fluor-4-methoxybenzoylchlorid
3-Fluor-4-methoxybenzoylchlorid
Formel: C₈H₆ClFO₂
Molecular Weight: 188.59 g/mol
MDL: MFCD03428589
CAS-Nummer: 3907-15-1
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR