2-((Difluormethyl)thio)benzoylchlorid

Lieferant: CHEMPUR

FL5770.5 FL5770.10
CHMPFL5770.5EA 0 EUR
CHMPFL5770.5 CHMPFL5770.10
2-((Difluormethyl)thio)benzoylchlorid
2-((Difluormethyl)thio)benzoylchlorid
Formel: C₈H₅ClF₂OS
Molecular Weight: 222,64 g/mol
Siedepunkt: 135…136 °C (1 mmHg)
MDL: MFCD01631519
CAS-Nummer: 79676-60-1
UN: 3265
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR