2,3,4,5-Tetrafluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 2,3,4,5-Tetrafluorbenzoesäureamid

005842-5G 005842-10G
FLUO005842-5GEA 0 EUR
FLUO005842-5G FLUO005842-10G
2,3,4,5-Tetrafluorbenzamid
2,3,4,5-Tetrafluorbenzamid
Formel: C₇H₃F₄NO
MDL: MFCD01631457
CAS-Nummer: 16582-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR