2,3,4-Trifluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 2,3,4-Trifluorbenzoesäureamid

007479-1G 007479-5G 007479-10G
FLUO007479-1GEA 45.9 EUR
FLUO007479-1G FLUO007479-5G FLUO007479-10G
2,3,4-Trifluorbenzamid
2,3,4-Trifluorbenzamid
Formel: C₇H₄F₃NO
Molecular Weight: 175.11 g/mol
MDL: MFCD00061265
CAS-Nummer: 207919-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR