2,3,5-Trifluorbenzamid

Lieferant: FLUOROCHEM

007955-5G 007955-10G 007955-1G
FLUO007955-5GEA 195 EUR
FLUO007955-5G FLUO007955-10G FLUO007955-1G
2,3,5-Trifluorbenzamid
2,3,5-Trifluorbenzamid
Formel: C₇H₄F₃NO
Molecular Weight: 175.11 g/mol
MDL: MFCD00061198
CAS-Nummer: 238403-46-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR