2,3,6-Trifluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 2,3,6-Trifluorbenzoesäureamid

008711-5G 008711-10G
FLUO008711-5GEA 82.1 EUR
FLUO008711-5G FLUO008711-10G
2,3,6-Trifluorbenzamid
2,3,6-Trifluorbenzamid
Formel: C₇H₄F₃NO
MDL: MFCD00061266
CAS-Nummer: 207986-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR