2,3-Difluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 2,3-Difluorbenzoesäureamid

004777-25G 004777-5G
FLUO004777-25GEA 0 EUR
FLUO004777-25G FLUO004777-5G
2,3-Difluorbenzamid
2,3-Difluorbenzamid
Formel: C₇H₅F₂NO
Molecular Weight: 157,12 g/mol
MDL: MFCD00061137
CAS-Nummer: 18355-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR