2,4,5-Trifluorbenzamid

Lieferant: FLUOROCHEM

007480-5G 007480-1G
FLUO007480-5GEA 152 EUR
FLUO007480-5G FLUO007480-1G
2,4,5-Trifluorbenzamid
2,4,5-Trifluorbenzamid
Formel: C₇H₄F₃NO
MDL: MFCD00082478
CAS-Nummer: 98349-23-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR