2,4,6-Trifluorbenzamid
Lieferant: FLUOROCHEM
007481-1G
007481-5G
FLUO007481-1GEA
36.9
EUR
FLUO007481-1G
FLUO007481-5G
2,4,6-Trifluorbenzamid
2,4,6-Trifluorbenzamid
Formel:
C₇H₄F₃NO |
MDL:
MFCD00061199 CAS-Nummer: 82019-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...