2,4-Difluorbenzamid
Lieferant: Thermo Scientific
Synonyme:
2,4-Difluorbenzoesäureamid
TL00450.1
TL00450.10
TL00450.50
MAYBTL00450.1EA
0
EUR
MAYBTL00450.1
MAYBTL00450.10
MAYBTL00450.50
2,4-Difluorbenzamid
2,4-Difluorbenzamid
Formel:
C₇H₅F₂NO Molecular Weight: 157,12 g/mol |
MDL:
MFCD00015547 CAS-Nummer: 85118-02-1 EINECS: 285-654-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...