2,5-Difluorbenzamid
Lieferant: FLUOROCHEM
Synonyme:
2,5-Difluorbenzoesäureamid
001933-10G
001933-25G
FLUO001933-10GEA
60.9
EUR
FLUO001933-10G
FLUO001933-25G
2,5-Difluorbenzamid
2,5-Difluorbenzamid
Formel:
C₇H₅F₂NO Molecular Weight: 157.12 g/mol |
MDL:
MFCD00015548 CAS-Nummer: 85118-03-2 EINECS: 285-655-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...