2,6-Difluorbenzamid
Lieferant: Thermo Scientific
Synonyme:
2,6-Difluorbenzoesäureamid
BTB06491.1
BTB06491.10
BTB06491.50
MAYBBTB06491.1EA
0
EUR
MAYBBTB06491.1
MAYBBTB06491.10
MAYBBTB06491.50
2,6-Difluorbenzamid
2,6-Difluorbenzamid
Formel:
C₇H₅F₂NO Molecular Weight: 157,12 g/mol Siedepunkt: 284 °C (1013 hPa) Schmelzpunkt: 144…145,5 °C |
MDL:
MFCD00007972 CAS-Nummer: 18063-03-1 EINECS: 241-972-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...