2,6-Difluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 2,6-Difluorbenzoesäureamid

001956-250G 001956-2.5KG 001956-100G
FLUO001956-250GEA 0 EUR
FLUO001956-250G FLUO001956-2.5KG FLUO001956-100G
2,6-Difluorbenzamid
2,6-Difluorbenzamid
Formel: C₇H₅F₂NO
Molecular Weight: 157.12 g/mol
Siedepunkt: 284
Schmelzpunkt: 144…145.5 °C
MDL: MFCD00007972
CAS-Nummer: 18063-03-1
EINECS: 241-972-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR