2-Fluorbenzamid
Lieferant: FLUOROCHEM
Synonyme:
2-Fluorbenzoesäureamid
002696-25G
002696-10G
FLUO002696-25GEA
48.6
EUR
FLUO002696-25G
FLUO002696-10G
2-Fluorbenzamid
2-Fluorbenzamid
Formel:
C₇H₆FNO Molecular Weight: 139.13 g/mol Siedepunkt: 287 Schmelzpunkt: 115…119 °C |
MDL:
MFCD00007970 CAS-Nummer: 445-28-3 EINECS: 207-157-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...