3,4,5-Trifluorbenzamid

Lieferant: FLUOROCHEM

007482-5G 007482-1G
FLUO007482-5GEA 171 EUR
FLUO007482-5G FLUO007482-1G
3,4,5-Trifluorbenzamid
3,4,5-Trifluorbenzamid
Formel: C₇H₄F₃NO
Molecular Weight: 175,11 g/mol
MDL: MFCD00083524
CAS-Nummer: 78324-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR