3,4-Difluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 3,4-Difluorbenzoesäureamid

004778-10G 004778-25G 004778-100G
FLUO004778-10GEA 0 EUR
FLUO004778-10G FLUO004778-25G FLUO004778-100G
3,4-Difluorbenzamid
3,4-Difluorbenzamid
Formel: C₇H₅F₂NO
Molecular Weight: 157,12 g/mol
MDL: MFCD00015549
CAS-Nummer: 85118-04-3
EINECS: 285-656-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR