3,5-Difluorbenzamid

Lieferant: FLUOROCHEM
Achtung

Synonyme: 3,5-Difluorbenzoesäureamid

004779-25G 004779-5G
FLUO004779-25GEA 159 EUR
FLUO004779-25G FLUO004779-5G
3,5-Difluorbenzamid
3,5-Difluorbenzamid
Formel: C₇H₅F₂NO
Molecular Weight: 157.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00061138
CAS-Nummer: 132980-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR