3-Fluorbenzamid

Lieferant: FLUOROCHEM

Synonyme: 3-Fluorbenzoesäureamid

006180-10G 006180-50G
FLUO006180-10GEA 49.2 EUR
FLUO006180-10G FLUO006180-50G
3-Fluorbenzamid
3-Fluorbenzamid
Formel: C₇H₆FNO
Molecular Weight: 139.13 g/mol
Siedepunkt: 287
Schmelzpunkt: 133
MDL: MFCD00007983
CAS-Nummer: 455-37-8
EINECS: 207-247-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR