4-Fluorbenzamid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Fluorbenzoesäureamid
197900050
197900250
ACRO197900050EA
32.8
EUR
ACRO197900050
ACRO197900250
4-Fluorbenzamid 98%
4-Fluorbenzamid
Formel:
C₇H₆FNO Molecular Weight: 139.13 g/mol Siedepunkt: 287 Schmelzpunkt: 154…158 °C |
MDL:
MFCD00007992 CAS-Nummer: 824-75-9 EINECS: 212-535-9 |
Spezifikation Testergebnisse
Appearance | White to beige to light brown Powder |
Infrared spectrum | Conforms |
Melting point | 154 to 158 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...