4-Fluorbenzamid
Lieferant: FLUOROCHEM
Synonyme:
4-Fluorbenzoesäureamid
006181-25G
006181-100G
FLUO006181-25GEA
52.1
EUR
FLUO006181-25G
FLUO006181-100G
4-Fluorbenzamid
4-Fluorbenzamid
Formel:
C₇H₆FNO Molecular Weight: 139.13 g/mol Siedepunkt: 287 Schmelzpunkt: 155 Dichte: 1.43 g/cm³ (20 °C) |
MDL:
MFCD00007992 CAS-Nummer: 824-75-9 EINECS: 212-535-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...