Pentafluorbenzamid
Lieferant: FLUOROCHEM
006947-10G
006947-25G
FLUO006947-10GEA
0
EUR
FLUO006947-10G
FLUO006947-25G
Pentafluorbenzamid
Pentafluorbenzamid
Formel:
C₇H₂F₅NO Siedepunkt: 272 °C (1013 hPa) Schmelzpunkt: 151 °C |
MDL:
MFCD00007971 CAS-Nummer: 652-31-3 EINECS: 211-488-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...