2-(Trifluormethoxy)benzamid

Lieferant: CHEMPUR

Synonyme: 2-(Trifluormethoxy)benzoesäureamid

TL00395.1 TL00395.10 TL00395.50
CHMPTL00395.1EA 0 EUR
CHMPTL00395.1 CHMPTL00395.10 CHMPTL00395.50
2-(Trifluormethoxy)benzamid
2-(Trifluormethoxy)benzamid
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
MDL: MFCD00042407
CAS-Nummer: 127979-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR