4-(Trifluormethoxy)benzamid
Lieferant: Thermo Scientific
Synonyme:
4-(Trifluormethoxy)benzoesäureamid
TL00456.1
TL00456.10
TL00456.50
MAYBTL00456.1EA
0
EUR
MAYBTL00456.1
MAYBTL00456.10
MAYBTL00456.50
4-(Trifluormethoxy)benzamid
4-(Trifluormethoxy)benzamid
Formel:
C₈H₆F₃NO₂ Molecular Weight: 205,14 g/mol |
MDL:
MFCD00041384 CAS-Nummer: 456-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...