4-(Trifluormethoxy)benzamid

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluormethoxy)benzoesäureamid

001543-1G 001543-5G 001543-25G
FLUO001543-1GEA 0 EUR
FLUO001543-1G FLUO001543-5G FLUO001543-25G
4-(Trifluormethoxy)benzamid
4-(Trifluormethoxy)benzamid
Formel: C₈H₆F₃NO₂
Molecular Weight: 205,14 g/mol
MDL: MFCD00041384
CAS-Nummer: 456-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR