2,4-Difluorbenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 2,4-Difluorbenzoesäurehydrazid, 2,4-Difluorbenzol-1-carbohydrazid, 2,4-Difluorbenzohydrazid

008285-1G 008285-5G
FLUO008285-1GEA 42.9 EUR
FLUO008285-1G FLUO008285-5G
2,4-Difluorbenzhydrazid
2,4-Difluorbenzhydrazid
Formel: C₇H₆F₂N₂O
Molecular Weight: 172,13 g/mol
MDL: MFCD00173685
CAS-Nummer: 118737-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR