3,4-Difluorbenzhydrazid
Lieferant: FLUOROCHEM
009137-1G
009137-5G
FLUO009137-1GEA
82.1
EUR
FLUO009137-1G
FLUO009137-5G
3,4-Difluorbenzhydrazid
3,4-Difluorbenzhydrazid
Formel:
C₇H₆F₂N₂O Molecular Weight: 172,13 g/mol |
MDL:
MFCD00672992 CAS-Nummer: 229957-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...