3,5-Difluorbenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 3,5-Difluorbenzohydrazid, 3,5-Difluorbenzoesäurehydrazid, 3,5-Difluorbenzol-1-carbohydrazid

009138-1G 009138-5G
FLUO009138-1GEA 69.8 EUR
FLUO009138-1G FLUO009138-5G
3,5-Difluorbenzhydrazid
3,5-Difluorbenzhydrazid
Formel: C₇H₆F₂N₂O
Molecular Weight: 172,13 g/mol
MDL: MFCD01310810
CAS-Nummer: 244022-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR