Methyl-2-fluorbenzoat

Lieferant: FLUOROCHEM

Synonyme: 2-Fluorbenzoesäuremethylester

006801-25G 006801-100G 006801-250G
FLUO006801-25GEA 42.9 EUR
FLUO006801-25G FLUO006801-100G FLUO006801-250G
Methyl-2-fluorbenzoat
Methyl-2-fluorbenzoat
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00017913
CAS-Nummer: 394-35-4
EINECS: 206-894-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR