Methyl-4-fluorbenzoat
Lieferant: FLUOROCHEM
Synonyme:
4-Fluorbenzoesäuremethylester
006802-25G
006802-100G
FLUO006802-25GEA
34.1
EUR
FLUO006802-25G
FLUO006802-100G
Methyl-4-fluorbenzoat
Methyl-4-fluorbenzoat
Formel:
C₈H₇FO₂ Molecular Weight: 154,14 g/mol |
MDL:
MFCD00017959 CAS-Nummer: 403-33-8 EINECS: 206-956-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...