Methyl-4-nitrobenzoat ≥99%
Lieferant: Thermo Scientific
Synonyme:
4-Nitrobenzoesäuremethylester
A12928.22
A12928.36
A12928.22EA
43.6
EUR
A12928.22
A12928.36
Methyl-4-nitrobenzoat ≥99%
Methyl-4-nitrobenzoat
Purity: 99%
CAS No.: 619-50-1
Molecular Formula: C8H7NO4
Formula Weight: 181.15
Melting Point: 95-97°
Beilstein Reference: 1912198
Formel:
C₈H₇NO₄ Molecular Weight: 181,15 g/mol Schmelzpunkt: 95…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007350 CAS-Nummer: 619-50-1 EINECS: 210-599-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...