2,3,6-Trifluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

007484-5G 007484-25G
FLUO007484-5GEA 57.9 EUR
FLUO007484-5G FLUO007484-25G
2,3,6-Trifluorbenzoesäure
2,3,6-Trifluorobenzoic acid
Formel: C₇H₃F₃O₂
Molecular Weight: 176,09 g/mol
Siedepunkt: 237 °C (1013 hPa)
Schmelzpunkt: 125…126 °C
MDL: MFCD00002409
CAS-Nummer: 2358-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR