4-Amino-2-fluorbenzoesäure

Lieferant: Apollo Scientific

PC9751-5G PC9751-25G PC9751-100G
APOSPC9751-5GEA 21.9 EUR
APOSPC9751-5G APOSPC9751-25G APOSPC9751-100G
4-Amino-2-fluorbenzoesäure
4-Amino-2-fluorobenzoic acid
Formel: C₇H₆FNO₂
Molecular Weight: 155.13 g/mol
MDL: MFCD01569397
CAS-Nummer: 446-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR