4-Amino-2-fluorbenzoesäure

Lieferant: BIONET RESEARCH

8T-0213.500 8T-0213.1 8T-0213.5 8T-0213.10 8T-0213.50
BIOT8T-0213.500EA 0 EUR
BIOT8T-0213.500 BIOT8T-0213.1 BIOT8T-0213.5 BIOT8T-0213.10 BIOT8T-0213.50
4-Amino-2-fluorbenzoesäure
4-Amino-2-fluorobenzoic acid
Formel: C₇H₆FNO₂
Molecular Weight: 155.13 g/mol
MDL: MFCD01569397
CAS-Nummer: 446-31-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR