4-Amino-3-(trifluormethoxy)benzoesäure
Lieferant: Thermo Scientific
TL00679.250
TL00679DA
MAYBTL00679.250EA
0
EUR
MAYBTL00679.250
MAYBTL00679DA
4-Amino-3-(trifluormethoxy)benzoesäure
4-Amino-3-(trifluoromethoxy)benzoic acid
Formel:
C₈H₆F₃NO₃ |
MDL:
MFCD00205143 CAS-Nummer: 175278-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...