3-Fluor-4-nitrobenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H26168.03 H26168.06
H26168.03EA 18.7 EUR
H26168.03 H26168.06
3-Fluor-4-nitrobenzoesäure ≥98%
3-Fluor-4-nitrobenzoesäure

Purity: 98+% CAS No.: 403-21-4 Molecular Formula: C7H4FNO4 Formula Weight: 185.11 Melting Point: 173-176° MDL No.: MFCD01862092

Formel: C₇H₄FNO₄
Schmelzpunkt: 173…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01862092
CAS-Nummer: 403-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR