3-Fluor-4-nitrobenzoesäure

Lieferant: BIONET RESEARCH
Achtung

11P-237.500 11P-237.1
BIOT11P-237.500EA 0 EUR
BIOT11P-237.500 BIOT11P-237.1
3-Fluor-4-nitrobenzoesäure
3-Fluor-4-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185.11 g/mol
MDL: MFCD01862092
CAS-Nummer: 403-21-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR