2,3-Difluor-4-ethoxybenzoesäure

Lieferant: CHEMPUR

FL13020.POR 013020-1G
CHMPFL13020.POREA 0 EUR
CHMPFL13020.POR FLUO013020-1G
2,3-Difluor-4-ethoxybenzoesäure
2,3-Difluoro-4-ethoxybenzoic acid
Formel: C₉H₈F₂O₃
Molecular Weight: 202,16 g/mol
MDL: MFCD02258849
CAS-Nummer: 124728-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR