2-(Trifluormethoxy)benzoesäure

Lieferant: FLUOROCHEM
Achtung

002256-1G 002256-5G 002256-25G
FLUO002256-1GEA 31.1 EUR
FLUO002256-1G FLUO002256-5G FLUO002256-25G
2-(Trifluormethoxy)benzoesäure
2-(Trifluormethoxy)benzoesäure
Formel: C₈H₅F₃O₃
Molecular Weight: 206,12 g/mol
MDL: MFCD00052325
CAS-Nummer: 1979-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR