2-Fluor-5-methoxybenzoesäure

Lieferant: CHEMPUR

CFF1121.1 FL13327.POR 013327-1G
CHMPCFF1121.1EA 522 EUR
CHMPCFF1121.1 CHMPFL13327.POR FLUO013327-1G
2-Fluor-5-methoxybenzoesäure
2-Fluoro-5-methoxybenzoic acid
Formel: C₈H₇FO₃
MDL: MFCD00272566
CAS-Nummer: 367-83-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR