3,4-Difluor-2-methoxybenzoesäure

Lieferant: CHEMPUR

FL13819.POR 013819-1G
CHMPFL13819.POREA 0 EUR
CHMPFL13819.POR FLUO013819-1G
3,4-Difluor-2-methoxybenzoesäure
3,4-Difluoro-2-methoxybenzoic acid
Formel: C₈H₆F₂O₃
MDL: MFCD03428593
CAS-Nummer: 875664-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR